Assay Details
| Gene Symbol: F7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080136.1 | NP_001073605.1 | 477 | 57 | ||
| GenBank mRNA | EF061407.2 | - | 477 | 57 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F7| Gene Symbol: F7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080136.1 | NP_001073605.1 | 477 | 57 | ||
| GenBank mRNA | EF061407.2 | - | 477 | 57 | ||
Set Membership: |
|