Assay Details
| Gene Symbol: PRSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047120.1 | NP_001040585.1 | 201 | 157 | ||
| XM_015135091.2 | XP_014990577.2 | 432 | 157 | |||
| XM_015135092.2 | XP_014990578.1 | 532 | 157 | |||
| XM_015135093.2 | XP_014990579.1 | 292 | 157 | |||
| XM_015135094.2 | XP_014990580.1 | 329 | 157 | |||
More Information
Set Membership: |
|

PRSS2
Genomic Map