Assay Details
| Gene Symbol: LILRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040672.2 | NP_001035762.2 | 2404 | 98 | ||
| XM_028839147.1 | XP_028694980.1 | 2860 | 98 | |||
| GenBank mRNA | DQ155431.1 | - | 2405 | 98 | ||
More Information
Set Membership: |
|

LILRB1
Genomic Map