Assay Details
| Gene Symbol: IFIT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261730.2 | NP_001248659.1 | 149 | 66 | ||
| GenBank mRNA | JU322796.1 | - | 167 | 66 | ||
| JV043641.1 | - | 149 | 66 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001261730.2 transcripts.
Set Membership: |
|

IFIT2
Genomic Map