Assay Details
| Gene Symbol: KLRC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104593.3 | NP_001098063.3 | 308 | 85 | ||
| GenBank mRNA | AF190934.1 | - | 85 | |||
| AF190936.1 | - | 85 | ||||
| AF190941.1 | - | 85 | ||||
| AF190942.1 | - | 85 | ||||
| AF294888.1 | - | 85 | ||||
| AF395616.1 | - | 85 | ||||
| JU486209.1 | - | 85 | ||||
More Information
Set Membership: |
|

KLRC3
Genomic Map