Assay Details
| Gene Symbol: SELENOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159414.2 | NP_001152886.1 | 552 | 79 | ||
| GenBank mRNA | JU332110.1 | - | 535 | 79 | ||
| JU470706.1 | - | 553 | 79 | |||
| JV703836.1 | - | 540 | 79 | |||

SELENOT
Genomic Map