Assay Details
| Gene Symbol: CCDC43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001114705.4 | XP_001114705.3 | 844 | 103 | ||
| XM_015119878.2 | XP_014975364.2 | 844 | 103 | |||
| GenBank mRNA | JU332892.1 | - | 229 | 103 | ||
| JU338659.1 | - | 229 | 103 | |||
More Information
Set Membership: |
|

CCDC43
Genomic Map