Assay Details
| Gene Symbol: ACTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258072.1 | NP_001245001.1 | 671 | 63 | ||
| GenBank mRNA | JU473021.1 | - | 671 | 63 | ||
| JV480232.1 | - | 664 | 63 | |||
| JV635492.1 | - | 702 | 63 | |||
More Information
Set Membership: |
|

ACTA2
Genomic Map