Assay Details
| Gene Symbol: MIR21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031992.1 | 32 | 64 | |||
| Gene Symbol: VMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119970.2 | XP_014975456.1 | 3227 | 64 | ||
| XM_015119971.2 | XP_014975457.1 | 2778 | 64 | |||
| XM_028835613.1 | XP_028691446.1 | 2755 | 64 | |||
| XM_028835614.1 | XP_028691447.1 | 2647 | 64 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
|---|
More Information
Set Membership: |
|

MIR21
Genomic Map