Assay Details
| Gene Symbol: ELFN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193792.2 | NP_001180721.1 | 2449 | 77 | ||
| XM_015150269.2 | XP_015005755.1 | 3415 | 77 | |||
| XM_015150270.2 | XP_015005756.1 | 3436 | 77 | |||
| XM_015150272.2 | XP_015005758.1 | 8502 | 77 | |||
| XM_015150273.2 | XP_015005759.1 | 3264 | 77 | |||
| XM_015150274.2 | XP_015005760.1 | 3285 | 77 | |||
| XM_028827199.1 | XP_028683032.1 | 8523 | 77 | |||
| XM_028827200.1 | XP_028683033.1 | 8160 | 77 | |||
| XM_028827201.1 | XP_028683034.1 | 8351 | 77 | |||
| XM_028827202.1 | XP_028683035.1 | 4451 | 77 | |||
| XM_028827203.1 | XP_028683036.1 | 8421 | 77 | |||
| GenBank mRNA | JV605045.1 | - | 3189 | 77 | ||
| JV667337.1 | - | 3148 | 77 | |||
More Information
Set Membership: |
|

ELFN2
Genomic Map