Assay Details
| Gene Symbol: METTL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193830.2 | NP_001180759.1 | 559 | 61 | ||
| XM_028846658.1 | XP_028702491.1 | 686 | 61 | |||
| GenBank mRNA | JU330569.1 | - | 220 | 61 | ||
More Information
Set Membership: |
|

METTL24
Genomic Map