Assay Details
| Gene Symbol: GDPGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194046.1 | NP_001180975.1 | 45 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194046.1 transcripts.
Set Membership: |
|

GDPGP1
Genomic Map