Assay Details
| Gene Symbol: CLN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194050.2 | NP_001180979.1 | 52 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194050.2 transcripts.
Set Membership: |
|

CLN8
Genomic Map