Assay Details
| Gene Symbol: COX6B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193383.1 | NP_001180312.1 | 216 | 82 | ||
| XM_015124938.2 | XP_014980424.1 | 322 | 82 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

COX6B2| Gene Symbol: COX6B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193383.1 | NP_001180312.1 | 216 | 82 | ||
| XM_015124938.2 | XP_014980424.1 | 322 | 82 | |||
Set Membership: |
|