Assay Details
| Gene Symbol: TMEM174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194141.2 | NP_001181070.1 | 660 | 61 | ||
| XM_015140196.2 | XP_014995682.1 | 729 | 61 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM174| Gene Symbol: TMEM174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194141.2 | NP_001181070.1 | 660 | 61 | ||
| XM_015140196.2 | XP_014995682.1 | 729 | 61 | |||
Set Membership: |
|