Assay Details
| Gene Symbol: NEURL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194339.3 | NP_001181268.1 | 1040 | 63 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEURL2| Gene Symbol: NEURL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194339.3 | NP_001181268.1 | 1040 | 63 | ||
Set Membership: |
|