Assay Details
| Gene Symbol: PAQR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194357.2 | NP_001181286.1 | 4746 | 103 | ||
| XM_015136468.2 | XP_014991954.1 | 4583 | 103 | |||
More Information
Set Membership: |
|

PAQR8
Genomic Map