Assay Details
| Gene Symbol: COX4I2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194480.2 | NP_001181409.1 | 419 | 63 | ||
| XM_028827357.1 | XP_028683190.1 | 393 | 63 | |||
| GenBank mRNA | JV635837.1 | - | 402 | 63 | ||
More Information
Set Membership: |
|

COX4I2
Genomic Map