Assay Details
| Gene Symbol: EEF1AKMT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194759.3 | NP_001181688.1 | 304 | 114 | ||
| GenBank mRNA | JV045090.1 | - | 230 | 114 | ||
| JV636087.1 | - | 268 | 114 | |||
More Information
Set Membership: |
|

EEF1AKMT3
Genomic Map