Assay Details
| Gene Symbol: TIMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194847.3 | NP_001181776.1 | 865 | 70 | ||
| GenBank mRNA | JV722213.1 | - | 300 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TIMP4| Gene Symbol: TIMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194847.3 | NP_001181776.1 | 865 | 70 | ||
| GenBank mRNA | JV722213.1 | - | 300 | 70 | ||
Set Membership: |
|