Assay Details
| Gene Symbol: ERV3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190831.1 | NP_001177760.1 | 752 | 110 | ||
| XM_015133398.2 | XP_014988884.1 | 1338 | 110 | |||
| GenBank mRNA | JU470886.1 | - | 1278 | 110 | ||
| JU470887.1 | - | 1278 | 110 | |||
| JV043953.1 | - | 1297 | 110 | |||
More Information
Set Membership: |
|

ERV3-1
Genomic Map