Assay Details
| Gene Symbol: WDR77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195443.1 | NP_001182372.1 | 516 | 99 | ||
| XM_028852444.1 | XP_028708277.1 | 914 | 99 | |||
| GenBank mRNA | CN648359.1 | - | 516 | 99 | ||
| JU336347.1 | - | 586 | 99 | |||
| JU477170.1 | - | 580 | 99 | |||
| JV048223.1 | - | 592 | 99 | |||
| JV638733.1 | - | 606 | 99 | |||
More Information
Set Membership: |
|

WDR77
Genomic Map