Assay Details
| Gene Symbol: PEBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190951.2 | NP_001177880.2 | 402 | 86 | ||
| XM_015144897.2 | XP_015000383.2 | 528 | 86 | |||
| XM_015144898.2 | XP_015000384.2 | 548 | 86 | |||
| GenBank mRNA | JV643578.1 | - | 453 | 86 | ||
More Information
Set Membership: |
|

PEBP4
Genomic Map