Assay Details
| Gene Symbol: LYRM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193351.1 | NP_001180280.1 | 125 | 76 | ||
| XM_015118865.2 | XP_014974351.1 | 220 | 76 | |||
| XM_028835696.1 | XP_028691529.1 | 218 | 76 | |||
| XM_028835697.1 | XP_028691530.1 | 284 | 76 | |||
| GenBank mRNA | JU333311.1 | - | 155 | 76 | ||
| JV045549.1 | - | 127 | 76 | |||
| JV459626.1 | - | 50 | 76 | |||
More Information
Set Membership: |
|

LYRM9
Genomic Map