Assay Details
| Gene Symbol: HSP90AB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195533.1 | NP_001182462.1 | 453 | 81 | ||
| XM_015136375.2 | XP_014991861.1 | 527 | 81 | |||
| XM_015136376.2 | XP_014991862.1 | 1815 | 81 | |||
| GenBank mRNA | CB230592.1 | - | 456 | 81 | ||
| JV504958.1 | - | 445 | 81 | |||
| JV667161.1 | - | 445 | 81 | |||
More Information
Set Membership: |
|

HSP90AB1
Genomic Map