Assay Details
| Gene Symbol: PTRHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193681.1 | NP_001180610.1 | 255 | 88 | ||
| GenBank mRNA | JV544619.1 | - | 60 | 88 | ||
| JV616402.1 | - | 261 | 88 | |||
| JV635542.1 | - | 260 | 88 | |||
More Information
Set Membership: |
|

PTRHD1
Genomic Map