Assay Details
| Gene Symbol: TNFAIP8L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193682.2 | NP_001180611.1 | 108 | 62 | ||
| GenBank mRNA | JV533214.1 | - | 105 | 62 | ||
| JV567836.1 | - | 73 | 62 | |||
| JV667295.1 | - | 76 | 62 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193682.2 transcripts.
Set Membership: |
|

TNFAIP8L1
Genomic Map