Assay Details
| Gene Symbol: SERTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193731.1 | NP_001180660.1 | 252 | 109 | ||
| XM_015120881.2 | XP_014976367.1 | 696 | 109 | |||
| XM_028837082.1 | XP_028692915.1 | 816 | 109 | |||
| XM_028837083.1 | XP_028692916.1 | 4040 | 109 | |||
| GenBank mRNA | JU322437.1 | - | 505 | 109 | ||
| JV602454.1 | - | 639 | 109 | |||
More Information
Set Membership: |
|

SERTM1
Genomic Map