Assay Details
| Gene Symbol: H2AFZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193550.1 | NP_001180479.1 | 65 | 72 | ||
| GenBank mRNA | CN643635.1 | - | 65 | 72 | ||
| JU319711.1 | - | 183 | 72 | |||
| JU471434.1 | - | 176 | 72 | |||
| JV044028.1 | - | 187 | 72 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193550.1 transcripts.
Set Membership: |
|

H2AFZ
Genomic Map