Assay Details
| Gene Symbol: LGALS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168627.1 | NP_001162098.1 | 319 | 94 | ||
| GenBank mRNA | EU152916.1 | - | 319 | 94 | ||
| JV044013.1 | - | 344 | 94 | |||
More Information
Set Membership: |
|

LGALS1
Genomic Map