Assay Details
| Gene Symbol: C11H12orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194760.3 | NP_001181689.3 | 831 | 113 | ||
| XM_001116886.4 | XP_001116886.3 | 1948 | 113 | |||
| XM_015152168.2 | XP_015007654.2 | 1949 | 113 | |||
More Information
Set Membership: |
|

C11H12orf56
Genomic Map