Assay Details
| Gene Symbol: C11H12orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194760.3 | NP_001181689.3 | 944 | 97 | ||
| XM_001116886.4 | XP_001116886.3 | 2061 | 97 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C11H12orf56| Gene Symbol: C11H12orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194760.3 | NP_001181689.3 | 944 | 97 | ||
| XM_001116886.4 | XP_001116886.3 | 2061 | 97 | |||