Assay Details
| Gene Symbol: C11H12orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194760.3 | NP_001181689.3 | 1031 | 95 | ||
| XM_001116886.4 | XP_001116886.3 | 2148 | 95 | |||
| XM_028829022.1 | XP_028684855.1 | 1829 | 95 | |||

C11H12orf56
Genomic Map