Assay Details
| Gene Symbol: TMEM263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194764.2 | NP_001181693.1 | 274 | 101 | ||
| GenBank mRNA | JU337001.1 | - | 74 | 101 | ||
| JV048632.1 | - | 75 | 101 | |||
| JV726469.1 | - | 74 | 101 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194764.2 transcripts.
Set Membership: |
|

TMEM263
Genomic Map