Assay Details
| Gene Symbol: DTYMK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194775.2 | NP_001181704.1 | 490 | 70 | ||
| XM_001094132.4 | XP_001094132.2 | 732 | 70 | |||
| XM_015111561.2 | XP_014967047.1 | 601 | 70 | |||
| XM_028830471.1 | XP_028686304.1 | 545 | 70 | |||
| GenBank mRNA | JV053794.1 | - | 471 | 70 | ||
More Information
Set Membership: |
|

DTYMK
Genomic Map