Assay Details
| Gene Symbol: RPL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193561.1 | NP_001180490.1 | 1146 | 96 | ||
| GenBank mRNA | CN805627.1 | - | 430 | 96 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPL15| Gene Symbol: RPL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193561.1 | NP_001180490.1 | 1146 | 96 | ||
| GenBank mRNA | CN805627.1 | - | 430 | 96 | ||
Set Membership: |
|