Assay Details
| Gene Symbol: UBB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194808.1 | NP_001181737.1 | 847 | 97 | ||
| XM_001087919.3 | XP_001087919.1 | 997 | 97 | |||
| XM_015118664.2 | XP_014974150.1 | 1207 | 97 | |||
| GenBank mRNA | JV627679.1 | - | 477 | 97 | ||
More Information
Set Membership: |
|

UBB
Genomic Map