Assay Details
| Gene Symbol: ATP5MC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198734.1 | NP_001185663.1 | 750 | 85 | ||
| GenBank mRNA | CN648600.1 | - | 560 | 85 | ||
| JU334974.1 | - | 813 | 85 | |||
| JU475987.1 | - | 827 | 85 | |||
| JU475989.1 | - | 905 | 85 | |||
More Information
Set Membership: |
|

ATP5MC3
Genomic Map