Assay Details
Gene Symbol: MIR365-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032489.1 | 40 | 84 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cytoplasmic polyadenylated homeobox like | CPHXL | -151.2kb | Sense | Chr. 20 - 62698937 - 62705863 on NCBI Build Mmul_10 | |
Downstream | contactin associated protein family member 4 | CNTNAP4 | 384.4kb | Anti-sense | Chr. 20 - 63242274 - 63522729 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
