Assay Details
| Gene Symbol: CHPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244592.2 | NP_001231521.1 | 265 | 89 | ||
| GenBank mRNA | CO579439.1 | - | 153 | 89 | ||
| JU476085.1 | - | 732 | 89 | |||
| JV584995.1 | - | 743 | 89 | |||
| JV654185.1 | - | 747 | 89 | |||
More Information
Set Membership: |
|

CHPF2
Genomic Map