Assay Details
| Gene Symbol: HNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195823.2 | NP_001182752.1 | 617 | 96 | ||
| XM_015110016.2 | XP_014965502.2 | 1813 | 96 | |||
| GenBank mRNA | CB309464.1 | - | 501 | 96 | ||
More Information
Set Membership: |
|

HNMT
Genomic Map