Assay Details
| Gene Symbol: VWA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260638.1 | NP_001247567.1 | 177 | 111 | ||
| GenBank mRNA | JU335962.1 | - | 177 | 111 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VWA1| Gene Symbol: VWA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260638.1 | NP_001247567.1 | 177 | 111 | ||
| GenBank mRNA | JU335962.1 | - | 177 | 111 | ||
Set Membership: |
|