Assay Details
| Gene Symbol: GOLGA7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260720.1 | NP_001247649.1 | 344 | 70 | ||
| GenBank mRNA | JU320919.1 | - | 344 | 70 | ||
| JV600360.1 | - | 381 | 70 | |||
More Information
Set Membership: |
|

GOLGA7B
Genomic Map