Assay Details
| Gene Symbol: GTF2E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260889.1 | NP_001247818.1 | 970 | 86 | ||
| XM_028843353.1 | XP_028699186.1 | 984 | 86 | |||
| GenBank mRNA | JU321059.1 | - | 989 | 86 | ||
| JU473756.1 | - | 971 | 86 | |||
| JU473757.1 | - | 971 | 86 | |||
| JV043413.1 | - | 970 | 86 | |||
More Information
Set Membership: |
|

GTF2E1
Genomic Map