Assay Details
| Gene Symbol: SNAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260982.1 | NP_001247911.1 | 777 | 84 | ||
| GenBank mRNA | JU472747.1 | - | 670 | 84 | ||
| JV047987.1 | - | 777 | 84 | |||
| JV636326.1 | - | 773 | 84 | |||
More Information
Set Membership: |
|

SNAI2
Genomic Map