Assay Details
| Gene Symbol: PTPN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261109.1 | NP_001248038.1 | 1999 | 98 | ||
| XM_015152836.2 | XP_015008322.1 | 2070 | 98 | |||
| GenBank mRNA | JU335687.1 | - | 2018 | 98 | ||
| JU335688.1 | - | 2006 | 98 | |||
| JU472515.1 | - | 2023 | 98 | |||
| JV045239.1 | - | 1999 | 98 | |||
| JV567459.1 | - | 2034 | 98 | |||
More Information
Set Membership: |
|

PTPN11
Genomic Map