Assay Details
| Gene Symbol: DHRS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261687.1 | NP_001248616.1 | 766 | 66 | ||
| XM_015143765.2 | XP_014999251.1 | 890 | 66 | |||
| GenBank mRNA | JU323169.1 | - | 766 | 66 | ||
| JU475949.1 | - | 708 | 66 | |||
| JV047338.1 | - | 756 | 66 | |||
| JV489166.1 | - | 739 | 66 | |||
More Information
Set Membership: |
|

DHRS7
Genomic Map