Assay Details
| Gene Symbol: DNAJC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261745.1 | NP_001248674.1 | 354 | 117 | ||
| XM_015114513.2 | XP_014969999.1 | 392 | 117 | |||
| XM_028832527.1 | XP_028688360.1 | 392 | 117 | |||
| GenBank mRNA | JU323308.1 | - | 342 | 117 | ||
| JV046157.1 | - | 354 | 117 | |||
More Information
Set Membership: |
|

DNAJC24
Genomic Map