Assay Details
| Gene Symbol: NUDT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261782.2 | NP_001248711.1 | 474 | 78 | ||
| XM_015138743.2 | XP_014994229.1 | 347 | 78 | |||
| XM_015138745.2 | XP_014994231.1 | 283 | 78 | |||
| GenBank mRNA | JU323329.1 | - | 531 | 78 | ||
| JU476026.1 | - | 552 | 78 | |||
| JV044417.1 | - | 474 | 78 | |||
| JV565138.1 | - | 639 | 78 | |||
More Information
Set Membership: |
|

NUDT9
Genomic Map