Assay Details
| Gene Symbol: ATP5MD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265824.1 | NP_001252753.1 | 102 | 108 | ||
| XM_015147965.2 | XP_015003451.1 | 293 | 108 | |||
| XM_015147966.2 | XP_015003452.1 | 285 | 108 | |||
| GenBank mRNA | JV489109.1 | - | 102 | 108 | ||
More Information
Set Membership: |
|

ATP5MD
Genomic Map